Gene Transcripts

Gene Symbol
ERAP2
Entrez Gene ID
64167
Gene Name
endoplasmic reticulum aminopeptidase 2
Gene Aliases
L-RAP, LRAP
Chromosome Location
Chr. 5: 96875940 - 96919703 on Build GRCh38
UniGene ID
Hs.482910
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011543544.1 XP_011541846.1 3 877
GenBank mRNA AB109031.1 - 3 844
AB163917.1 - 3 844
AF191545.1 - 2 816
AK074281.1 - 3 921
AK312864.1 - 3 857
AY028805.1 - 2 774
BC017927.2 - 2 817
BC065240.1 - 3 838 6166289

View Full Product Details