Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001001396.2 | NP_001001396.1 | 22 | 4872 | |
NM_001684.4 | NP_001675.3 | 21 | 4694 | ||
GenBank mRNA | AK307104.1 | - | 5 | 1017 | |
BC140774.1 | - | 22 | 4544 | 9021642 | |
BX537444.1 | - | 21 | 4921 | ||
BX537745.1 | - | 1 | 1339 | ||
BX647701.1 | - | 4 | 650 | ||
CR749393.1 | - | 21 | 4624 | ||
CR936708.1 | - | 21 | 4649 | ||
M25874.1 | - | 20 | 4194 | ||
M83363.1 | - | 21 | 4372 |