Gene Transcripts

Gene Symbol
MEOX1
Entrez Gene ID
4222
Gene Name
mesenchyme homeobox 1
Gene Aliases
KFS2, MOX1
Chromosome Location
Chr. 17: 43640389 - 43661954 on Build GRCh38
UniGene ID
Hs.438
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040002.1 NP_001035091.1 4 1475
NM_004527.3 NP_004518.1 3 1621
NM_013999.3 NP_054705.1 2 1448
GenBank mRNA BG677020.1 - 1 432
BX284090.1 - 1 432
U10492.1 - 3 1622
U10493.1 - 2 1449

View Full Product Details