Gene Transcripts

Gene Symbol
SERPINE2
Entrez Gene ID
5270
Gene Name
serpin family E member 2
Gene Aliases
GDN, GDNPF, PI-7, PI7, PN-1, PN1, PNI
Chromosome Location
Chr. 2: 223975048 - 224039319 on Build GRCh38
UniGene ID
Hs.38449
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136528.1 NP_001130000.1 3, 4 733
NM_001136530.1 NP_001130002.1 3, 4 663
NM_006216.3 NP_006207.1 3, 4 733
NR_073116.1 4, 5 1151
XM_005246641.2 XP_005246698.1 3, 4 822
XM_017004329.1 XP_016859818.1 3, 4 714
XM_017004330.1 XP_016859819.1 3, 4 551
XM_017004331.1 XP_016859820.1 3, 4 714
XM_017004332.1 XP_016859821.1 3, 4 551
GenBank mRNA A03911.1 - 2, 3 514
AK225823.1 - 3, 4 701
AK295564.1 - 3, 4 631
AK297588.1 - 3, 4 538
AK312499.1 - 3, 4 747
BC015663.2 - 3, 4 690 4578406
BC042628.1 - 4, 5 1150 4824856
M17783.1 - 2, 3 490

View Full Product Details