Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001001975.1 | NP_001001975.1 | 4 | 565 | |
NM_001687.4 | NP_001678.1 | 4 | 565 | ||
GenBank mRNA | BC002389.2 | - | 4 | 565 | |
BC004426.1 | - | 4 | 565 | 2819749 | |
BC018079.1 | - | 4 | 467 | 4798666 | |
BC018817.2 | - | 4 | 565 | ||
CR542218.1 | - | 4 | 464 | ||
S87916.1 | - | 4 | 547 | ||
S87918.1 | - | 4 | 547 | ||
X63422.1 | - | 4 | 547 | ||
X63423.1 | - | 4 | 547 |