Gene Transcripts

Gene Symbol
CYB561D1
Entrez Gene ID
284613
Gene Name
cytochrome b561 family member D1
Gene Aliases
Chromosome Location
Chr. 1: 109492830 - 109500441 on Build GRCh38
UniGene ID
Hs.514682
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134400.1 NP_001127872.1 3 624
NM_001134402.1 NP_001127874.1 2 430
NM_001134403.1 NP_001127875.1 4 662
NM_001134404.1 NP_001127876.1 4 739
NM_182580.2 NP_872386.1 3 558
XM_005270775.2 XP_005270832.1 3 635
XM_005270776.3 XP_005270833.1 3 474
XM_005270777.2 XP_005270834.1 2 514
XM_011541286.2 XP_011539588.1 4 623
XM_011541287.2 XP_011539589.2 3 1751
XM_017001078.1 XP_016856567.1 4 546
XM_017001079.1 XP_016856568.1 3 368
GenBank mRNA AK096354.1 - 3 537
AK126707.1 - 3 635
AK128798.1 - 4 703
AK294992.1 - 3 589
AK299086.1 - 3 424
AK303546.1 - 2 499
BC093683.1 - 3 532 7939528
BC111999.1 - 3 532 8327467
BC143308.1 - 2 494
BX647509.1 - 4 642
DA054191.1 - 2 414
DC376706.1 - 3 468

View Full Product Details