Gene Transcripts

Gene Symbol
CSTF2T
Entrez Gene ID
23283
Gene Name
cleavage stimulation factor subunit 2, tau variant
Gene Aliases
CstF-64T
Chromosome Location
Chr. 10: 51695486 - 51699595 on Build GRCh38
UniGene ID
Hs.591358
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015235.2 NP_056050.1 1 2193
GenBank mRNA AB014589.1 - 1 1941
AK222574.1 - 1 2179
AL136747.1 - 1 2189
AY130299.1 - 1 2184
BC028239.1 - 1 2176 5244025

View Full Product Details