Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001293640.1 | NP_001280569.1 | 9 | 668 | |
NM_015275.2 | NP_056090.1 | 9 | 668 | ||
XM_011538073.2 | XP_011536375.1 | 9 | 668 | ||
XM_011538074.2 | XP_011536376.1 | 9 | 634 | ||
XM_011538077.2 | XP_011536379.1 | 9 | 668 | ||
XM_017019082.1 | XP_016874571.1 | 9 | 668 | ||
GenBank mRNA | AB028956.1 | - | 9 | 650 | |
AK001657.1 | - | 9 | 668 | ||
AK001728.1 | - | 9 | 633 | ||
AK023146.1 | - | 6 | 303 | ||
AL137753.1 | - | 6 | 291 | ||
BC104992.1 | - | 9 | 610 | 8143995 | |
BC104994.1 | - | 9 | 610 | 8143997 | |
BC110850.1 | - | 9 | 619 | ||
BC143373.1 | - | 9 | 610 | 9051878 |