Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024953.3 | NP_079229.2 | 12 | 1331 | |
XM_005253938.1 | XP_005253995.1 | 11 | 1177 | ||
XM_006719606.2 | XP_006719669.1 | 12 | 1991 | ||
XM_017019977.1 | XP_016875466.1 | 10 | 1047 | ||
GenBank mRNA | AB054990.1 | - | 12 | 1273 | |
AK023151.1 | - | 9 | 976 | ||
AK292485.1 | - | 12 | 1304 | ||
AK307494.1 | - | 13 | 1480 | ||
AK309772.1 | - | 11 | 1157 | ||
BC034357.1 | - | 12 | 1285 | ||
BC065481.1 | - | 12 | 1264 | ||
BC113585.1 | - | 12 | 1331 | 8322637 | |
BC113587.1 | - | 12 | 1331 | 8322639 | |
BX641140.1 | - | 12 | 1286 | ||
BX647450.1 | - | 12 | 1284 | ||
BX647992.1 | - | 9 | 932 |