Gene Transcripts

Gene Symbol
Sfrp4
Entrez Gene ID
20379
Gene Name
secreted frizzled-related protein 4
Gene Aliases
Chromosome Location
Chr. 13: 19623175 - 19632825 on Build GRCm38
UniGene ID
Mm.42095
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016687.3 NP_057896.1 2 735
GenBank mRNA AF117709.1 - 2 750
AK080766.1 - 2 736
BC034853.1 - 2 694 1364770

View Full Product Details