Gene Transcripts

Gene Symbol
Sntg2
Entrez Gene ID
268534
Gene Name
syntrophin, gamma 2
Gene Aliases
2210008K22Rik, 9530013L23, BB121248
Chromosome Location
Chr. 12: 30174480 - 30373392 on Build GRCm38
UniGene ID
Mm.279765
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172951.3 NP_766539.2 9 767
GenBank mRNA AK008673.1 - 9 675
AK035310.1 - 9 721
AK087560.1 - 9 717
AK137345.1 - 9 726

View Full Product Details