Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008743.2 | NP_032769.2 | 6 | 873 | |
GenBank mRNA | AB006812.1 | - | 6 | 819 | |
AK033701.1 | - | 6 | 1881 | ||
AV377313.2 | - | 4 | 461 | ||
BC138852.1 | - | 6 | 836 | 8861874 | |
BC138855.1 | - | 6 | 836 | 8861877 | |
BC145443.1 | - | 6 | 745 | ||
CJ211561.1 | - | 3 | 193 | ||
Y09688.1 | - | 6 | 845 |