Gene Transcripts

Gene Symbol
Chrnb2
Entrez Gene ID
11444
Gene Name
cholinergic receptor, nicotinic, beta polypeptide 2 (neuronal)
Gene Aliases
Acrb-2, Acrb2, C030030P04Rik, [b]2-nAchR
Chromosome Location
Chr. 3: 89753448 - 89764632 on Build GRCm38
UniGene ID
Mm.35088
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009602.4 NP_033732.2 6 1925
GenBank mRNA AK049722.1 - 6 1898
AK051742.1 - 6 1907
BC065103.1 - 6 1878 6838257

View Full Product Details