Gene Transcripts

Gene Symbol
Olfr862
Entrez Gene ID
258555
Gene Name
olfactory receptor 862
Gene Aliases
MOR146-1
Chromosome Location
Chr. 9: 19883383 - 19884303 on Build GRCm38
UniGene ID
Mm.434360
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146562.1 NP_666773.1 1 268
GenBank mRNA BC120769.1 - 1 320
BC125340.1 - 1 320 40129855

View Full Product Details