Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001102420.2 | NP_001095890.1 | 7 | 1514 | |
NM_001102421.2 | NP_001095891.1 | 6 | 1377 | ||
NM_001278243.1 | NP_001265172.1 | 7 | 1093 | ||
NM_001278244.1 | NP_001265173.1 | 6 | 956 | ||
NM_001278245.1 | NP_001265174.1 | 7 | 1090 | ||
NM_006007.3 | NP_005998.1 | 6 | 1374 | ||
GenBank mRNA | AF062346.1 | - | 6 | 1097 | |
AF062347.1 | - | 6 | 952 | ||
AK025782.1 | - | 7 | 977 | ||
AK290849.1 | - | 6 | 1097 | ||
AK307376.1 | - | 4 | 798 | ||
BC011018.1 | - | 6 | 1213 | 4331933 | |
BC073131.1 | - | 6 | 878 | 6137219 | |
BX648551.1 | - | 1 | 4305 | ||
BX649046.1 | - | 1 | 902 |