Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030709.2 | NP_109634.2 | 8 | 790 | |
GenBank mRNA | AB016229.1 | - | 8 | 788 | |
AB016230.1 | - | 8 | 790 | ||
AB016423.1 | - | 4 | 405 | ||
AB041037.1 | - | 8 | 747 | ||
AK029714.1 | - | 9 | 1637 | ||
BC132151.1 | - | 7 | 736 | 40130428 | |
BC145027.1 | - | 7 | 718 | 9053575 |