Gene Transcripts

Gene Symbol
Acbd6
Entrez Gene ID
72482
Gene Name
acyl-Coenzyme A binding domain containing 6
Gene Aliases
0610010G04Rik, 2610100E10Rik
Chromosome Location
Chr. 1: 155558098 - 155687316 on Build GRCm38
UniGene ID
Mm.288249
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145781.1 NP_001139253.1 1, 2 745
NM_026683.1 NP_080959.1 1, 2 745
NM_028250.3 NP_082526.2 1, 2 745
GenBank mRNA AK002470.1 - 1, 2 746
AK011781.1 - 1, 2 738
AK082937.1 - 1, 2 746
AK165836.1 - 1, 2 740
AK167959.1 - 2, 3 686
AK209754.1 - 1, 2 60
BC028537.1 - 1, 2 490
BC061029.1 - 1, 2 471
BC115580.1 - 1, 2 337 40102241
BC115581.1 - 1, 2 337 40102242

View Full Product Details