Gene Transcripts
- Gene Symbol
- Fam177a
- Entrez Gene ID
- 73385
- Gene Name
- family with sequence similarity 177, member A
- Gene Aliases
- 1700047I17Rik, 1700047I17Rik1, C14orf24, D730001C10Rik, Fam177a1
- Chromosome Location
- Chr. 12: 55124468 - 55142104 on Build GRCm38
- UniGene ID
- Mm.439875
- Species
- Mus musculus
- Gene Symbol
- 1700047I17Rik2
- Entrez Gene ID
- 100101807
- Gene Name
- RIKEN cDNA 1700047I17 gene 2
- Gene Aliases
- Chromosome Location
- Chr. 12: 55199532 - 55217168 on Build GRCm38
- UniGene ID
- Mm.439875
- Species
- Mus musculus
View Full Product Details