Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019447.2 | NP_062320.2 | 6 | 709 | |
GenBank mRNA | AF099017.1 | - | 6 | 669 | |
AF224724.1 | - | 6 | 669 | ||
AK007987.1 | - | 6 | 707 | ||
AK075718.1 | - | 6 | 618 | ||
AK144554.1 | - | 6 | 681 | ||
AK149522.1 | - | 6 | 673 | ||
AK165433.1 | - | 5 | 827 | ||
AK169989.1 | - | 6 | 705 | ||
BC019376.1 | - | 6 | 674 | 5134851 |