Gene Transcripts

Gene Symbol
Grin2d
Entrez Gene ID
24412
Gene Name
glutamate receptor, ionotropic, N-methyl D-aspartate 2D
Gene Aliases
GluN2D
Chromosome Location
Chr. 1: 101819026 - 101856881 on Build Rnor_6.0
UniGene ID
Rn.91209
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022797.1 NP_073634.1 13 4989
XM_008759317.1 XP_008757539.1 13 4613
GenBank mRNA L31611.1 - 12 4264
L31612.1 - 13 4989

View Full Product Details