Gene Transcripts

Gene Symbol
Cptp
Entrez Gene ID
313771
Gene Name
ceramide-1-phosphate transfer protein
Gene Aliases
Gltpd1
Chromosome Location
Chr. 5: 173314228 - 173318384 on Build Rnor_6.0
UniGene ID
Rn.3197
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007703.1 NP_001007704.1 3 1102
XM_006239576.2 XP_006239638.1 3 1263
GenBank mRNA BC083599.1 - 3 1102

View Full Product Details