Gene Transcripts

Gene Symbol
Fam207a
Entrez Gene ID
294333
Gene Name
family with sequence similarity 207, member A
Gene Aliases
RGD1311257
Chromosome Location
Chr. 20: 11863145 - 11896833 on Build Rnor_6.0
UniGene ID
Rn.22122
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008307.1 NP_001008308.1 6 906
GenBank mRNA BC083697.1 - 6 906

View Full Product Details