Gene Transcripts

Gene Symbol
Isyna1
Entrez Gene ID
290651
Gene Name
inositol-3-phosphate synthase 1
Gene Aliases
Chromosome Location
Chr. 16: 20589497 - 20592333 on Build Rnor_6.0
UniGene ID
Rn.98850
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013880.2 NP_001013902.2 10 1556
GenBank mRNA BC079011.1 - 8 1560
FQ229915.1 - 5 852

View Full Product Details