Gene Transcripts

Gene Symbol
Olr1129
Entrez Gene ID
405178
Gene Name
olfactory receptor 1129
Gene Aliases
Chromosome Location
Chr. 8: 18992899 - 18993861 on Build Rnor_6.0
UniGene ID
Rn.143607
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000879.1 NP_001000879.1 1 909

View Full Product Details