Gene Transcripts

Gene Symbol
Aph1a
Entrez Gene ID
365872
Gene Name
APH1A gamma secretase subunit
Gene Aliases
RGD1311546
Chromosome Location
Chr. 2: 198006269 - 198010992 on Build Rnor_6.0
UniGene ID
Rn.22786
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014255.1 NP_001014277.1 4, 5 681
XM_006232977.2 XP_006233039.1 4, 5 734
GenBank mRNA BC087081.1 - 4, 5 681

View Full Product Details