Gene Transcripts

Gene Symbol
Capn13
Entrez Gene ID
362701
Gene Name
calpain 13
Gene Aliases
CANP 13
Chromosome Location
Chr. 6: 24579590 - 24676533 on Build Rnor_6.0
UniGene ID
Rn.15541
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025133.1 NP_001020304.1 23 2205
XM_006239777.2 XP_006239839.1 24 2363
XM_006239778.2 XP_006239840.1 23 2316
XM_006239779.2 XP_006239841.1 24 2359
GenBank mRNA BC091250.1 - 23 2205

View Full Product Details