Gene Transcripts

Gene Symbol
GABRR2
Entrez Gene ID
2570
Gene Name
gamma-aminobutyric acid type A receptor rho2 subunit
Gene Aliases
Chromosome Location
Chr. 6: 89254464 - 89315299 on Build GRCh38
UniGene ID
Hs.379330
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002043.4 NP_002034.3 2 298
GenBank mRNA BC130352.1 - 2 217
BC130354.1 - 2 217 40146385
BX490975.1 - 2 306
M86868.1 - 2 244

View Full Product Details