Gene Transcripts

Gene Symbol
Rnase10
Entrez Gene ID
75019
Gene Name
ribonuclease, RNase A family, 10 (non-active)
Gene Aliases
4930474F22Rik, Rah1
Chromosome Location
Chr. 14: 51007751 - 51010761 on Build GRCm38
UniGene ID
Mm.318775
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162863.1 NP_001156335.1 2 791
NM_029145.3 NP_083421.2 2 900
GenBank mRNA AK015573.1 - 2 792
AY226990.1 - 2 900

View Full Product Details