Gene Transcripts

Gene Symbol
GABRA1
Entrez Gene ID
2554
Gene Name
gamma-aminobutyric acid type A receptor alpha1 subunit
Gene Aliases
ECA4, EIEE19, EJM, EJM5
Chromosome Location
Chr. 5: 161847191 - 161899959 on Build GRCh38
UniGene ID
Hs.175934
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000806.5 NP_000797.2 4, 5 654
NM_001127643.1 NP_001121115.1 4, 5 541
NM_001127644.1 NP_001121116.1 3, 4 690
NM_001127645.1 NP_001121117.1 3, 4 278
NM_001127648.1 NP_001121120.1 2, 3 300
GenBank mRNA AK223447.1 - 3, 4 427
AK289792.1 - 4, 5 654
BC030696.1 - 3, 4 255 4820972
DA228536.1 - 1, 2 55
DC307259.1 - 3, 4 278
DC325179.1 - 3, 4 502
DC334834.1 - 2, 3 300
R60404.1 - 1, 2 114
X13584.1 - 3, 4 446
X14766.1 - 3, 4 400

View Full Product Details