Gene Transcripts

Gene Symbol
FAM171A2
Entrez Gene ID
284069
Gene Name
family with sequence similarity 171 member A2
Gene Aliases
Chromosome Location
Chr. 17: 44353215 - 44363867 on Build GRCh38
UniGene ID
Hs.154001
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198475.2 NP_940877.2 2 293
GenBank mRNA BC029298.1 - 2 309
BC030200.1 - 2 293

View Full Product Details