Gene Transcripts

Gene Symbol
Zbed3
Entrez Gene ID
361881
Gene Name
zinc finger, BED-type containing 3
Gene Aliases
Chromosome Location
Chr. 2: 25039497 - 25052129 on Build Rnor_6.0
UniGene ID
Rn.103511
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025729.1 NP_001020900.1 2 1635
XM_006231816.2 XP_006231878.1 2 1777
GenBank mRNA BC097974.1 - 2 1635

View Full Product Details