Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145928.1 | NP_001139400.1 | 3 | 410 | |
NM_024545.3 | NP_078821.2 | 3 | 410 | ||
XM_005263767.3 | XP_005263824.1 | 3 | 384 | ||
XM_005263768.3 | XP_005263825.1 | 3 | 360 | ||
XM_005263769.3 | XP_005263826.1 | 3 | 281 | ||
XM_005263770.3 | XP_005263827.1 | 3 | 360 | ||
XM_006712748.3 | XP_006712811.1 | 3 | 570 | ||
XM_006712749.3 | XP_006712812.1 | 3 | 570 | ||
XM_017004903.1 | XP_016860392.1 | 3 | 360 | ||
XM_017004904.1 | XP_016860393.1 | 3 | 360 | ||
XM_017004905.1 | XP_016860394.1 | 3 | 360 | ||
GenBank mRNA | AK055979.1 | - | 3 | 326 | |
AK315185.1 | - | 3 | 410 | ||
AL136833.1 | - | 3 | 408 | ||
AY220791.1 | - | 3 | 278 | ||
BC117255.1 | - | 3 | 376 | 40125806 | |
BC143898.1 | - | 3 | 376 | 9052416 |