Gene Transcripts

Gene Symbol
HIST2H2AC
Entrez Gene ID
8338
Gene Name
histone cluster 2, H2ac
Gene Aliases
H2A, H2A-GL101, H2A/q, H2AFQ
Chromosome Location
Chr. 1: 149886975 - 149887411 on Build GRCh38
UniGene ID
Hs.408067
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003517.2 NP_003508.1 1 365
GenBank mRNA BC060324.1 - 1 400

View Full Product Details