Gene Transcripts

Gene Symbol
Atp13a3
Entrez Gene ID
224088
Gene Name
ATPase type 13A3
Gene Aliases
AU022875, Gm1745, Gm541, Gm542
Chromosome Location
Chr. 16: 30312423 - 30388856 on Build GRCm38
UniGene ID
Mm.8924
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128094.1 NP_001121566.1 33 6272
NM_001128096.1 NP_001121568.1 34 6362
GenBank mRNA AK145038.1 - 1 1678

View Full Product Details