Gene Transcripts

Gene Symbol
Ankub1
Entrez Gene ID
242037
Gene Name
ankrin repeat and ubiquitin domain containing 1
Gene Aliases
Gm410
Chromosome Location
Chr. 3: 57656493 - 57692780 on Build GRCm38
UniGene ID
Mm.291091
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033349.2 NP_001028521.1 4 952
GenBank mRNA AK134766.1 - 4 909
AK138483.1 - 4 952

View Full Product Details