Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022594.1 | NP_072116.1 | 3 | 346 | |
GenBank mRNA | BC060565.1 | - | 3 | 352 | |
BC062226.1 | - | 3 | 352 | 6890529 | |
FQ214121.1 | - | 3 | 348 | ||
FQ230435.1 | - | 3 | 359 | ||
U08976.1 | - | 3 | 346 |