Gene Transcripts

Gene Symbol
Stxbp3
Entrez Gene ID
114095
Gene Name
syntaxin binding protein 3
Gene Aliases
Munc18-c
Chromosome Location
Chr. 2: 211595763 - 211638756 on Build Rnor_6.0
UniGene ID
Rn.211988
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053637.2 NP_446089.1 6, 7 564
GenBank mRNA AB046544.1 - 6, 7 565
BC098660.1 - 6, 7 492 7386801

View Full Product Details