Gene Transcripts

Gene Symbol
Hoxb5
Entrez Gene ID
15413
Gene Name
homeobox B5
Gene Aliases
AI385717, Hox-2.1
Chromosome Location
Chr. 11: 96303512 - 96306121 on Build GRCm38
UniGene ID
Mm.207
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008268.2 NP_032294.2 2 1702
GenBank mRNA AK133512.1 - 2 1618
BC040755.1 - 3333
M26283.1 - 2 1702

View Full Product Details