Gene Transcripts

Gene Symbol
Kpna4
Entrez Gene ID
16649
Gene Name
karyopherin (importin) alpha 4
Gene Aliases
1110058D08Rik, IPOA3
Chromosome Location
Chr. 3: 69072221 - 69127092 on Build GRCm38
UniGene ID
Mm.30601
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008467.4 NP_032493.1 17 2329
GenBank mRNA AK147920.1 - 17 2338
BC026821.1 - 17 2124 3672708
BC052162.1 - 17 2330 6510795

View Full Product Details