Gene Transcripts

Gene Symbol
Sebox
Entrez Gene ID
18292
Gene Name
SEBOX homeobox
Gene Aliases
OG9, Og9x
Chromosome Location
Chr. 11: 78503513 - 78505081 on Build GRCm38
UniGene ID
Mm.134360
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008759.2 NP_032785.1 3 514
GenBank mRNA AK139838.1 - 3 538
AK143429.1 - 3 526
BC098184.1 - 3 514 5367903

View Full Product Details