Gene Transcripts

Gene Symbol
GRIN2D
Entrez Gene ID
2906
Gene Name
glutamate ionotropic receptor NMDA type subunit 2D
Gene Aliases
EB11, GluN2D, NMDAR2D, NR2D
Chromosome Location
Chr. 19: 48394875 - 48444937 on Build GRCh38
UniGene ID
Hs.445015
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000836.2 NP_000827.2 3 615
XM_011526872.1 XP_011525174.1 2 783
GenBank mRNA U77783.1 - 3 615

View Full Product Details