Gene Transcripts

Gene Symbol
Plxnc1
Entrez Gene ID
54712
Gene Name
plexin C1
Gene Aliases
2510048K12Rik, AW742158, CD232, Vespr
Chromosome Location
Chr. 10: 94790866 - 94945156 on Build GRCm38
UniGene ID
Mm.256712
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018797.2 NP_061267.1 2, 3 1211
GenBank mRNA AF190578.1 - 2, 3 1211
AK084603.1 - 2, 3 851

View Full Product Details