Gene Transcripts

Gene Symbol
CNGB3
Entrez Gene ID
54714
Gene Name
cyclic nucleotide gated channel beta 3
Gene Aliases
ACHM1
Chromosome Location
Chr. 8: 86573608 - 86743675 on Build GRCh38
UniGene ID
Hs.154433
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019098.4 NP_061971.3 2 198
GenBank mRNA AF272900.1 - 2 196
AL713036.1 - 2 199
BC150601.1 - 2 176 9056971

View Full Product Details