Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145924.1 | NP_001139396.1 | 3 | 803 | |
NM_001145925.1 | NP_001139397.1 | 3 | 664 | ||
NM_028137.3 | NP_082413.2 | 3 | 682 | ||
GenBank mRNA | AK011381.1 | - | 3 | 805 | |
AK013199.1 | - | 3 | 683 | ||
AK017795.1 | - | 3 | 807 | ||
AK075956.1 | - | 3 | 676 | ||
BC028651.1 | - | 2 | 205 | ||
BC052696.1 | - | 3 | 725 | 5715147 |