Gene Transcripts

Gene Symbol
Pnma3
Entrez Gene ID
245468
Gene Name
paraneoplastic antigen MA3
Gene Aliases
Chromosome Location
Chr. X: 73064787 - 73068191 on Build GRCm38
UniGene ID
Mm.256249
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153169.2 NP_694809.1 1 192
GenBank mRNA BC036726.1 - 1 230

View Full Product Details