Gene Transcripts

Gene Symbol
Ccdc65
Entrez Gene ID
105833
Gene Name
coiled-coil domain containing 65
Gene Aliases
4933417K04Rik, AI428139, AU020178, NYD-SP28
Chromosome Location
Chr. 15: 98708227 - 98723336 on Build GRCm38
UniGene ID
Mm.26205
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153518.1 NP_705738.1 3, 4 656
GenBank mRNA AF454432.1 - 3, 4 656
AK077174.1 - 3, 4 690
BC048073.1 - 3, 4 676 6312435

View Full Product Details