gRNA Design Details

Target DNA Sequence
TCAGTTATAAGACGGCGTAT
PAM Sequence
CGG
Target locus
Chr.15: 66119162 - 66119184 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 96.45/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
MEGF11
Entrez Gene ID
84465
Gene Name
multiple EGF like domains 11
Gene Aliases
-
Chromosome Location
Chr.15: 65895296 - 66253737 on GRCh38
UniGene ID
Hs.712886
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_032445.2 NP_115821.2 4
XM_017022670.1 XP_016878159.1 3
XM_017022671.1 XP_016878160.1 3
XM_017022672.1 XP_016878161.1 3
XM_017022673.1 XP_016878162.1 3