gRNA Design Details
- Target DNA Sequence
- CATAAGTGCACTCCGTGTCC
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 25326420 - 25326442 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.88/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Megf8
- Entrez Gene ID
-
269878
- Gene Name
- multiple EGF-like-domains 8
- Gene Aliases
-
AW049492, Egfl4, b2b1702Clo, b2b288Clo, m687Ddg, mKIAA0817
- Chromosome Location
-
Chr.7: 25316742 - 25365917 on GRCm38
- UniGene ID
- Mm.38527
- Species
-
Mouse