gRNA Design Details
- Target DNA Sequence
- CATCGCCAAAGACCGCTGTG
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 93256330 - 93256352 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.39/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Sned1
- Entrez Gene ID
-
208777
- Gene Name
- sushi, nidogen and EGF-like domains 1
- Gene Aliases
-
6720455I24Rik, AI197264, AI642697, D430044C15Rik, SST3, Snep
- Chromosome Location
-
Chr.1: 93235815 - 93301065 on GRCm38
- UniGene ID
- Mm.271897
- Species
-
Mouse