gRNA Design Details

Target DNA Sequence
TCACTCAGAGACAGCCAGAT
PAM Sequence
GGG
Target locus
Chr.6: 13215754 - 13215776 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 83.68/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Vwde
Entrez Gene ID
232585
Gene Name
von Willebrand factor D and EGF domains
Gene Aliases
Gm467
Chromosome Location
Chr.6: 13185610 - 13224965 on GRCm38
UniGene ID
Mm.389082
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001013757.2 NP_001013779.2 3